Learn More
Chemglass Life Sciences Vial, 10mL, Amber, 18mm Screw Thread Headspace, Flat Bottom, 22x46mm
SDP
Catalog No. 501532473
Encompass_Preferred
Vial, 10mL, Amber, 18mm Screw Thread Headspace, Flat Bottom, 22x46mm. These vials are manufactured from Type 1 Class A borosilicate glass. The radius bottom vials can be used on LEAP Technologies Inc, CTC Analytics, Varian, Gerstel and other instruments using CombiPAL and other models requiring magnetic capable closures.
Catalog No. 50-153-2473 Supplier Chemglass Life Sciences Supplier No. CV12942246
Vial, 10mL, Amber, 18mm Screw Thread Headspace, Flat Bottom, 22x46mm. These vials are manufactured from Type 1 Class A borosilicate glass. The radius bottom vials can be used on LEAP Technologies Inc, CTC Analytics, Varian, Gerstel and other instruments using CombiPAL and other models requiring magnetic capable closures.