Learn More
Waters Corp Waters Vial, Amber Glass, 12 x 32 mm, 2 mL
Catalog No. 50785985 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Amber Glass 12 x 32 mm Snap Neck Vial is designed to prevent the deterioration of delicate samples. It has a compact structure for a secure fit with analytical equipment and a stable composition to withstand certain chemical and mechanical stresses. The chemical inertness of the vial ensures no interference with analysis. The amber color prevents deterioration of delicate laboratory materials.
  • Compact structure provides a secure fit for analytical equipment.
  • Composition is stable enough to endure certain chemical and mechanical stresses.
  • Chemical inertness ensures no interference with analysis.
  • UV-tested by HPLC to ensure no contaminants.
  • Each batch is checked for cleanliness after packaging.
Catalog No. 50-785-985 Supplier Waters Corp Supplier No. WAT094220
The Amber Glass 12 x 32 mm Snap Neck Vial is designed to prevent the deterioration of delicate samples. It has a compact structure for a secure fit with analytical equipment and a stable composition to withstand certain chemical and mechanical stresses. The chemical inertness of the vial ensures no interference with analysis. The amber color prevents deterioration of delicate laboratory materials.
  • Compact structure provides a secure fit for analytical equipment.
  • Composition is stable enough to endure certain chemical and mechanical stresses.
  • Chemical inertness ensures no interference with analysis.
  • UV-tested by HPLC to ensure no contaminants.
  • Each batch is checked for cleanliness after packaging.