Learn More
Waters Corp Waters Crimp Vial, Amber Glass, 12 x 32 mm, 2 mL Volume
Catalog No. 50785989 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The amber glass material offers excellent protection against light, making it suitable for storing light-sensitive samples. These crimp vials are designed to be compatible with most common autosamplers, ensuring easy integration into laboratory workflows. While durable, they are recommended for single-use to maintain sample integrity and minimize contamination risk.
  • Provides excellent protection against light, making it suitable for storing light-sensitive samples.
  • Designed to be compatible with most common autosamplers.
  • Recommended for single-use to ensure sample integrity and minimize contamination risk.
  • Volume capacity of 2 mL.
  • Each pack contains 100 vials.
Catalog No. 50-785-989 Supplier Waters Corp Supplier No. WAT094223
The amber glass material offers excellent protection against light, making it suitable for storing light-sensitive samples. These crimp vials are designed to be compatible with most common autosamplers, ensuring easy integration into laboratory workflows. While durable, they are recommended for single-use to maintain sample integrity and minimize contamination risk.
  • Provides excellent protection against light, making it suitable for storing light-sensitive samples.
  • Designed to be compatible with most common autosamplers.
  • Recommended for single-use to ensure sample integrity and minimize contamination risk.
  • Volume capacity of 2 mL.
  • Each pack contains 100 vials.