Learn More
Waters Corp Waters Crimp Vial, Clear Glass, 12 x 32 mm, 2 mL
Catalog No. 50785987 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This clear glass crimp vial is designed for use with a vial crimper and can be sealed off for transportation of samples. It is suitable for sample preparation and collection, especially when sealing samples before shipping or transferring to another system. Using this vial with an accompanying aluminum crimp cap helps secure the sample for travel and protect it from contaminants. This crimp vial provides a long and durable sealing solution for your sample.
  • Designed for use with a vial crimper for sealing.
  • Can be sealed off for transportation of samples.
  • Comes in a pack of 100.
  • Strictly QC tested for cleanliness properties and dimensional tolerances.
  • Provides a long and durable sealing solution for samples.
  • Protects samples from contaminants.
Catalog No. 50-785-987 Supplier Waters Corp Supplier No. WAT094222
This clear glass crimp vial is designed for use with a vial crimper and can be sealed off for transportation of samples. It is suitable for sample preparation and collection, especially when sealing samples before shipping or transferring to another system. Using this vial with an accompanying aluminum crimp cap helps secure the sample for travel and protect it from contaminants. This crimp vial provides a long and durable sealing solution for your sample.
  • Designed for use with a vial crimper for sealing.
  • Can be sealed off for transportation of samples.
  • Comes in a pack of 100.
  • Strictly QC tested for cleanliness properties and dimensional tolerances.
  • Provides a long and durable sealing solution for samples.
  • Protects samples from contaminants.