Learn More
Med Vet International Green Prescription Vial, 16 Dram, 260/Case
Catalog No. 502643427
Encompass_Preferred
The Oasis 16 Dram child resistant prescription pill bottle is designed for storing medication. This prescription bottle comes in Amber and has a push down and turn cap making it difficult for children to open, but easy for the elderly. The light resistant vial is child resistant with 5% free easy open snap on caps. This product contains 270/case prescription vials. Height without cap: 8cm (3.15") Diameter 3.1cm (1.22"). EASY TO USE:  These bottles are easy to use and you can safely store your medicine or other small items in them. These prescription pharmacy pill bottles have push and turn caps. PERFECT TO ORGANIZE AND STORE PILLS:  These medicine containers are perfect to organize and store pills. The bottles can be used for pharmacies, physicians, labs, hospitals, and vets. LIGHTWEIGHT AND DURABLE PLASTIC:  These medicine bottles are made of premium quality plastic material.
Catalog No. 50-264-3427 Supplier Med Vet International Supplier No. VIAL16DG
The Oasis 16 Dram child resistant prescription pill bottle is designed for storing medication. This prescription bottle comes in Amber and has a push down and turn cap making it difficult for children to open, but easy for the elderly. The light resistant vial is child resistant with 5% free easy open snap on caps. This product contains 270/case prescription vials. Height without cap: 8cm (3.15") Diameter 3.1cm (1.22"). EASY TO USE:  These bottles are easy to use and you can safely store your medicine or other small items in them. These prescription pharmacy pill bottles have push and turn caps. PERFECT TO ORGANIZE AND STORE PILLS:  These medicine containers are perfect to organize and store pills. The bottles can be used for pharmacies, physicians, labs, hospitals, and vets. LIGHTWEIGHT AND DURABLE PLASTIC:  These medicine bottles are made of premium quality plastic material.