Learn More
TWD Scientific LLC Vial, Cryovial, Twd Biox, 1.5Ml, 1D And 2D Barcoded, External Thread, Blue Cap, Sterile, Writing Patch, Marked Graduations, Polypropylene, 1000 Vials/Case
SDP
Catalog No. 502285064 Shop All TWD Scientific LLC Products
Encompass_Preferred
1.5Ml Clear Polypropylene Sterile Pre-Set 1D & 2D Barcoded Cryovials With External Thread And Blue Caps Assembled. Cryovials Have Writing Patch And Marked Graduations. 25 Cryovials/Bag, 20 Bags/Pack, 2 Packs/Case, Price/Case
This item is not returnable. View return policy
Catalog No. 50-228-5064 Supplier TWD Scientific LLC Supplier No. BX889153
1.5Ml Clear Polypropylene Sterile Pre-Set 1D & 2D Barcoded Cryovials With External Thread And Blue Caps Assembled. Cryovials Have Writing Patch And Marked Graduations. 25 Cryovials/Bag, 20 Bags/Pack, 2 Packs/Case, Price/Case