Learn More
TWD Scientific LLC Vial, Cryovial, Twd Biox, 5.0Ml, 1D And 2D Barcoded, External Thread, White Cap, Sterile, Writing Patch, Marked Graduations, Polypropylene, 1000 Vials/Case
Shop All TWD Scientific LLC Products