Learn More
TWD Scientific LLC Vial, Cryovial, Twd Biox, 0.5Ml, 1D Barcoded, External Thread, White Cap, Sterile, Writing Patch, Marked Graduations, Polypropylene, 1000 Vials/Case
SDP
Catalog No. 502285047 Shop All TWD Scientific LLC Products
Encompass_Preferred
0.5Ml Clear Polypropylene Sterile Pre-Set 1D Barcoded Cryovials With External Thread And White Caps Assembled. Cryovials Have Writing Patch And Marked Graduations. 25 Cryovials/Bag, 20 Bags/Pack, 2 Packs/Case, Price/Case
This item is not returnable. View return policy
Catalog No. 50-228-5047 Supplier TWD Scientific LLC Supplier No. BX886050
0.5Ml Clear Polypropylene Sterile Pre-Set 1D Barcoded Cryovials With External Thread And White Caps Assembled. Cryovials Have Writing Patch And Marked Graduations. 25 Cryovials/Bag, 20 Bags/Pack, 2 Packs/Case, Price/Case