Learn More
TWD Scientific LLC BioX 1D Barcoded 2.0mL Cryovial, Purple Cap, Internal Thread. Case of 1000
SDP
Catalog No. 502274259 Shop All TWD Scientific LLC Products
Encompass_Preferred
BioX 2.0mL clear polypropylene sterile pre-set 1D barcoded cryovials with internal thread and purple caps assembled. Cryovials have a writing patch and marked graduations. 25 cryovials/bag, 20 bags/pack, 2 packs/case, price/case
This item is not returnable. View return policy
Catalog No. 50-227-4259 Supplier TWD Scientific LLC Supplier No. BX886215
BioX 2.0mL clear polypropylene sterile pre-set 1D barcoded cryovials with internal thread and purple caps assembled. Cryovials have a writing patch and marked graduations. 25 cryovials/bag, 20 bags/pack, 2 packs/case, price/case