Learn More
TWD Scientific LLC BioX Tricode 1.5mL Cryovial, Purple Cap, External Thread. Case of 1000
SDP
Catalog No. 502274244 Shop All TWD Scientific LLC Products
Encompass_Preferred
BioX 1.5mL clear polypropylene sterile pre-set 1D & 2D barcoded cryovials with external thread and purple caps assembled. Cryovials have writing patch and marked graduations. 25 cryovials/bag, 20 bags/pack, 2 packs/case, price/case
This item is not returnable. View return policy
Catalog No. 50-227-4244 Supplier TWD Scientific LLC Supplier No. BX889155
BioX 1.5mL clear polypropylene sterile pre-set 1D & 2D barcoded cryovials with external thread and purple caps assembled. Cryovials have writing patch and marked graduations. 25 cryovials/bag, 20 bags/pack, 2 packs/case, price/case