Learn More
Waters Corp Waters Vial, LCGC Certified, Amber Glass, 1 mL, Snap Neck
Catalog No. 50785205 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These certified vials are rigorously tested for cleanliness via HPLC and crafted to the industry’s most stringent dimensional tolerances. The tight construction ensures a secure fit with analytical equipment. Made from chemically and mechanically stable amber glass, they are also chemically inert, protecting samples from UV light deterioration and preventing any interference with your analysis.
  • Tight construction for secure fit with analytical equipment.
  • Chemically and mechanically stable and inert.
  • Amber glass protects samples from UV light deterioration.
  • UV-tested by HPLC; each batch checked for cleanliness.
  • Held to the industry’s tightest dimensional specifications.
Catalog No. 50-785-205 Supplier Waters Corp Supplier No. WAT025053C
These certified vials are rigorously tested for cleanliness via HPLC and crafted to the industry’s most stringent dimensional tolerances. The tight construction ensures a secure fit with analytical equipment. Made from chemically and mechanically stable amber glass, they are also chemically inert, protecting samples from UV light deterioration and preventing any interference with your analysis.
  • Tight construction for secure fit with analytical equipment.
  • Chemically and mechanically stable and inert.
  • Amber glass protects samples from UV light deterioration.
  • UV-tested by HPLC; each batch checked for cleanliness.
  • Held to the industry’s tightest dimensional specifications.