Learn More
Chemglass Life Sciences Vial, 2.0mL, Amber, Silanized, Large Opening, 12x32mm, GPI 10-425
SDP
Catalog No. 501532517
Encompass_Preferred
Vial, 2.0mL, Amber, Silanized, Large Opening, 12x32mm, GPI 10-425. With a 40% larger opening, the Large Opening Vial is specifically designed to eliminate broken needles due to the increased target area. These vials incorporate the unique Step Vial design that precisely centers a limited volume insert in the neck of the vial.
Catalog No. 50-153-2517 Supplier Chemglass Life Sciences Supplier No. CV26351232
Vial, 2.0mL, Amber, Silanized, Large Opening, 12x32mm, GPI 10-425. With a 40% larger opening, the Large Opening Vial is specifically designed to eliminate broken needles due to the increased target area. These vials incorporate the unique Step Vial design that precisely centers a limited volume insert in the neck of the vial.