Learn More
Chemglass Life Sciences Vial, 2.0mL, Amber with White Grad Spot, Snap Seal, 12x32mm, 11mm Snap Seal
SDP
Catalog No. 501532450
Encompass_Preferred
The Snap Seal™ vial can be securely sealed with the patented Poly Crimp™ Seal, aluminum seal or variety of Chem Snap Cap™ designs. The Snap Seal™ vial incorporates the unique Step Vial design that precisely centers a limited volume insert in the vial neck. This certifies dependable, reliable use with autosamplers, even those with robotic arms.
Catalog No. 50-153-2450 Supplier Chemglass Life Sciences Supplier No. CV11111232
The Snap Seal™ vial can be securely sealed with the patented Poly Crimp™ Seal, aluminum seal or variety of Chem Snap Cap™ designs. The Snap Seal™ vial incorporates the unique Step Vial design that precisely centers a limited volume insert in the vial neck. This certifies dependable, reliable use with autosamplers, even those with robotic arms.