Learn More
Chemglass Life Sciences Vial, 2.0mL, Amber with White Grad Spot, Standard Opening, 12x32mm, GPI 8-425
SDP
Catalog No. 50230732
Encompass_Preferred
Vial, 2.0mL, Amber with White Grad Spot, Standard Opening, 12x32mm, GPI 8-425. Standard opening screw thread vials contain a narrow opening allowing the use of GPI 8-425 closures and septa. This vial is designed to work in a variety of auto samplers requiring narrow neck vials. Available with graduated marking spots and silanized.
Catalog No. 50-230-732 Supplier Chemglass Life Sciences Supplier No. CV11201232
Vial, 2.0mL, Amber with White Grad Spot, Standard Opening, 12x32mm, GPI 8-425. Standard opening screw thread vials contain a narrow opening allowing the use of GPI 8-425 closures and septa. This vial is designed to work in a variety of auto samplers requiring narrow neck vials. Available with graduated marking spots and silanized.