Learn More
Chemglass Life Sciences Vial, 2.0mL, Amber with White Grad Spot, Large Opening, 12x32mm, 11mm Crimp
SDP
Catalog No. 501532459
Encompass_Preferred
Vial, 2.0mL, Amber with White Grad Spot, Large Opening, 12x32mm, 11mm Crimp. The 40% larger opening prevents broken needles due to the increased target area. The neck finish allows for the use of the patented Poly Crimp seals or standard aluminum seals. This vial is designed to work auto samples incorporating a robotic arm as well as other auto samplers. The vial incorporates the unique Step Vial design that precisely centers a limited volume insert in the vial neck.
Catalog No. 50-153-2459 Supplier Chemglass Life Sciences Supplier No. CV11881232
Vial, 2.0mL, Amber with White Grad Spot, Large Opening, 12x32mm, 11mm Crimp. The 40% larger opening prevents broken needles due to the increased target area. The neck finish allows for the use of the patented Poly Crimp seals or standard aluminum seals. This vial is designed to work auto samples incorporating a robotic arm as well as other auto samplers. The vial incorporates the unique Step Vial design that precisely centers a limited volume insert in the vial neck.