Learn More
Chemglass Life Sciences Vial, 2.0mL, Clear, Snap Seal, 12x32mm, 11mm Snap Seal
SDP
Catalog No. 501532449
Encompass_Preferred
Vial, 2.0mL, Clear, Snap Seal, 12x32mm, 11mm Snap Seal. The Snap Seal vial can be securely sealed with the patented Poly Crimp Seal, aluminum seal or variety of Chem Snap Cap designs. The Snap Seal vial incorporates the unique Step Vial design that precisely centers a limited volume insert in the vial neck. This certifies dependable, reliable use with autosamplers, even those with robotic arms.
Catalog No. 50-153-2449 Supplier Chemglass Life Sciences Supplier No. CV11081232
Vial, 2.0mL, Clear, Snap Seal, 12x32mm, 11mm Snap Seal. The Snap Seal vial can be securely sealed with the patented Poly Crimp Seal, aluminum seal or variety of Chem Snap Cap designs. The Snap Seal vial incorporates the unique Step Vial design that precisely centers a limited volume insert in the vial neck. This certifies dependable, reliable use with autosamplers, even those with robotic arms.