Learn More
Chemglass Life Sciences Vial, 2.0mL, Clear, Standard Opening, 12x32mm, 11mm Crimp
SDP
Catalog No. 501532456
Encompass_Preferred
Vial, 2.0mL, Clear, Standard Opening, 12x32mm, 11mm Crimp. The neck finish allows for the use of the patented Poly Crimp Seals or standard aluminum seals. This vial is designed to work auto samples incorporating a robotic arm as well as other auto samplers. The vial incorporates the unique Step Vial design that precisely centers a limited volume insert in the vial neck.
Catalog No. 50-153-2456 Supplier Chemglass Life Sciences Supplier No. CV11681232
Vial, 2.0mL, Clear, Standard Opening, 12x32mm, 11mm Crimp. The neck finish allows for the use of the patented Poly Crimp Seals or standard aluminum seals. This vial is designed to work auto samples incorporating a robotic arm as well as other auto samplers. The vial incorporates the unique Step Vial design that precisely centers a limited volume insert in the vial neck.