Learn More
Chemglass Life Sciences Vial, 2.0mL, Clear, Silanized, Large Opening, 12x32mm, 9mm Thread
SDP
Catalog No. 501532520
Encompass_Preferred
Vial, 2.0mL, Clear, Silanized, Large Opening, 12x32mm, 9mm Thread. With a 40% larger opening, these vials are specifically designed to work in robotic arm auto samplers. They also incorporate the unique Step Vial design that precisely centers a limited volume insert in the neck of the vial. Available with graduated marking spots, and silanized.
Catalog No. 50-153-2520 Supplier Chemglass Life Sciences Supplier No. CV26551232
Vial, 2.0mL, Clear, Silanized, Large Opening, 12x32mm, 9mm Thread. With a 40% larger opening, these vials are specifically designed to work in robotic arm auto samplers. They also incorporate the unique Step Vial design that precisely centers a limited volume insert in the neck of the vial. Available with graduated marking spots, and silanized.