Learn More
Chemglass Life Sciences Vial, 2.0mL, Clear with Marking Spot, Silanized, Standard Opening, 12x32mm, GPI 8-425
SDP
Catalog No. 501532511
Encompass_Preferred
Vial, 2.0mL, Clear with Marking Spot, Silanized, Standard Opening, 12x32mm, GPI 8-425. Standard opening screw thread vials contain a narrow opening allowing the use of GPI 8-425 closures and septa. This vial is designed to work in a variety of auto samplers requiring narrow neck vials.
Catalog No. 50-153-2511 Supplier Chemglass Life Sciences Supplier No. CV26001232
Vial, 2.0mL, Clear with Marking Spot, Silanized, Standard Opening, 12x32mm, GPI 8-425. Standard opening screw thread vials contain a narrow opening allowing the use of GPI 8-425 closures and septa. This vial is designed to work in a variety of auto samplers requiring narrow neck vials.