Learn More
Chemglass Life Sciences Vial, 2 Dram, 17 x 60 mm, 15-425 Thread, Pressure-Relief Cap (Pack of 48)
SDP
Catalog No. 502733524
Encompass_Preferred
These 2 dram borosilicate reaction vials are designed for use in heated and chemical reactions where internal pressure may develop. The vials include open-top pressure-relief caps to limit internal pressure and are supplied in convenient shelf packs for storage and handling.
  • Borosilicate glass construction.
  • 2 dram capacity.
  • 17 x 60 mm dimensions.
  • 15-425 thread compatibility.
  • Pressure-relief cap prevents pressure from exceeding 150 psig.
  • Supplied in shelf packs of 48 in SBS-coated trays with partitions.
Catalog No. 50-273-3524 Supplier Chemglass Life Sciences Supplier No. CG4912482
These 2 dram borosilicate reaction vials are designed for use in heated and chemical reactions where internal pressure may develop. The vials include open-top pressure-relief caps to limit internal pressure and are supplied in convenient shelf packs for storage and handling.
  • Borosilicate glass construction.
  • 2 dram capacity.
  • 17 x 60 mm dimensions.
  • 15-425 thread compatibility.
  • Pressure-relief cap prevents pressure from exceeding 150 psig.
  • Supplied in shelf packs of 48 in SBS-coated trays with partitions.