Learn More
Chemglass Life Sciences Chemglass Vial, 2 Dram, 17 x 60 mm, 15-425 Thread, Red Pressure-Relief Cap - Pack of 24
SDP
Catalog No. NC2595613
Encompass_Preferred
This 2 dram borosilicate reaction vial is designed for small-scale reactions and secure sample storage. It is supplied with a pressure-relief cap to help manage internal pressure during heated or pressurized procedures.
  • Capacity: 2 dram.
  • Dimensions: 17 x 60 mm.
  • Thread: 15-425 for standard closures.
  • Includes a red pressure-relief cap to reduce overpressure risk.
  • Made from borosilicate glass for chemical resistance.
  • Packaged 24 vials per shelf pack.
Catalog No. NC2595613 Supplier Chemglass Life Sciences Supplier No. CG4912242
This 2 dram borosilicate reaction vial is designed for small-scale reactions and secure sample storage. It is supplied with a pressure-relief cap to help manage internal pressure during heated or pressurized procedures.
  • Capacity: 2 dram.
  • Dimensions: 17 x 60 mm.
  • Thread: 15-425 for standard closures.
  • Includes a red pressure-relief cap to reduce overpressure risk.
  • Made from borosilicate glass for chemical resistance.
  • Packaged 24 vials per shelf pack.