Learn More
Chemglass Life Sciences Vial, 20mL, Clear, 20mm Headspace, Round Bottom, 23x75mm, 20mm Crimp, for use with Perkin Elmer Autosamplers
SDP
Catalog No. 501532471
Encompass_Preferred
Vial, 20mL, Clear, 20mm Headspace, Round Bottom, 23x75mm, 20mm Crimp, for use with Perkin Elmer Autosamplers. These vials are manufactured from Type 1 Class A Borosilicate glass. Please Note: Pressure Release Aluminum Seals are Designed to Open When the Internal Pressure Exceeds 3.0 +/- 0.5 Bar.
Catalog No. 50-153-2471 Supplier Chemglass Life Sciences Supplier No. CV12812375
Vial, 20mL, Clear, 20mm Headspace, Round Bottom, 23x75mm, 20mm Crimp, for use with Perkin Elmer Autosamplers. These vials are manufactured from Type 1 Class A Borosilicate glass. Please Note: Pressure Release Aluminum Seals are Designed to Open When the Internal Pressure Exceeds 3.0 +/- 0.5 Bar.