Learn More
Chemglass Life Sciences Vial, 20mL, Clear, 18mm Screw Thread Headspace, Flat Bottom w Large Radius Corners, 22x75mm
SDP
Catalog No. 501532472
Encompass_Preferred
These vials are manufactured from Type 1 Class A borosilicate glass. These flat bottom vials with large radius corners can be used on LEAP Technologies Inc, CTC Analytics, Varian, Gerstel and other instruments using CombiPAL and other models requiring magnetic capable closures.
Catalog No. 50-153-2472 Supplier Chemglass Life Sciences Supplier No. CV12922275
These vials are manufactured from Type 1 Class A borosilicate glass. These flat bottom vials with large radius corners can be used on LEAP Technologies Inc, CTC Analytics, Varian, Gerstel and other instruments using CombiPAL and other models requiring magnetic capable closures.