Learn More
Chemglass Life Sciences Chemglass Reaction Vial, 20 mL, 24-414 Thread, Red Pressure-Relief Cap, Pack of 24
SDP
Catalog No. NC2335756
Encompass_Preferred
Chemglass 20 mL reaction vials are made from type 1 borosilicate glass for use in heated reactions and pressure-relief applications. Each vial is supplied with a red pressure-relief screw cap and is sold in packs of 24.
  • 20 mL capacity.
  • 28 x 57 mm dimensions (diameter x height).
  • 24-414 screw thread.
  • Red pressure relief screw cap to help prevent overpressure.
  • Type 1 borosilicate glass construction.
  • Pack quantity: 24 vials.
Catalog No. NC2335756 Supplier Chemglass Life Sciences Supplier No. CG4912245
Chemglass 20 mL reaction vials are made from type 1 borosilicate glass for use in heated reactions and pressure-relief applications. Each vial is supplied with a red pressure-relief screw cap and is sold in packs of 24.
  • 20 mL capacity.
  • 28 x 57 mm dimensions (diameter x height).
  • 24-414 screw thread.
  • Red pressure relief screw cap to help prevent overpressure.
  • Type 1 borosilicate glass construction.
  • Pack quantity: 24 vials.