Learn More
Chemglass Life Sciences Vial, 28 x 70mm, 24-414 Thread, 30mL
SDP
Catalog No. 5000850164
Encompass_Preferred
Vial 30mL 28 x 70mm 24-414 Thread with Red Pressure Relief Cap Reaction vials are perfect for use in heated reactions Supplied complete with a special pressure relief open top cap Packed 100 per box Supplied complete with a special open top cap which prevents the pressure in the vial from exceeding 150 psig Made from 33 expansion borosilicate glass which conforms to ASTM E-438 Type 1-Class A USP Type 1 and ISO 3585 The high resistance to thermal shock along with its maximum chemical resistance makes this the glass of choice for reaction vials along with most all laboratory glassware Packaged in a SBS coated white tray complete with SBS partition Boxes are marked with alpha-numeric coordinates for fast and easy vial identification PLEASE NOTE VIALS AND CAPS ARE NOT AVAILABLE SEPARATE MUST BE PURCHASED COMPLETE
Catalog No. 50-008-50164 Supplier Chemglass Life Sciences Supplier No. CG491230
Vial 30mL 28 x 70mm 24-414 Thread with Red Pressure Relief Cap Reaction vials are perfect for use in heated reactions Supplied complete with a special pressure relief open top cap Packed 100 per box Supplied complete with a special open top cap which prevents the pressure in the vial from exceeding 150 psig Made from 33 expansion borosilicate glass which conforms to ASTM E-438 Type 1-Class A USP Type 1 and ISO 3585 The high resistance to thermal shock along with its maximum chemical resistance makes this the glass of choice for reaction vials along with most all laboratory glassware Packaged in a SBS coated white tray complete with SBS partition Boxes are marked with alpha-numeric coordinates for fast and easy vial identification PLEASE NOTE VIALS AND CAPS ARE NOT AVAILABLE SEPARATE MUST BE PURCHASED COMPLETE