Learn More
TWD Scientific LLC BioX 1.0mL SBS Format Minitube Combo. Case of 1920 Tubes and 20 Racks.
SDP
Catalog No. 502274266 Shop All TWD Scientific LLC Products
Encompass_Preferred
BioX 1.0ml SBS format screw top minitubes with blue caps. Vials come loaded into a barcoded 96-well rack with square wells to eliminate slip. The internally threaded cryovials feature 2D barcodes on the bottom and are gamma radiation sterilized. 96 tubes/rack, 20 racks/case. price/case.
This item is not returnable. View return policy
Catalog No. 50-227-4266 Supplier TWD Scientific LLC Supplier No. BX894213
BioX 1.0ml SBS format screw top minitubes with blue caps. Vials come loaded into a barcoded 96-well rack with square wells to eliminate slip. The internally threaded cryovials feature 2D barcodes on the bottom and are gamma radiation sterilized. 96 tubes/rack, 20 racks/case. price/case.