Learn More
Microsolv Technology Corporation Vials, Screw Top, Glass. Amber, 4ml, 13-425mm threads, 15x45mm outer dimensions. For use as an autosampler vial. 100/PK.
Catalog No. 5001452235
Encompass_Preferred
First hydrolytic, Class A, Borosilicate Glass, amberized. Features ND13 threads. Amber vials are often used for light sensitive compounds and considered low actinic.
Catalog No. 50-014-52235 Supplier Microsolv Technology Corporation Supplier No. 9504SE0AV
First hydrolytic, Class A, Borosilicate Glass, amberized. Features ND13 threads. Amber vials are often used for light sensitive compounds and considered low actinic.