Learn More
Chemglass Life Sciences Vial, 4.0mL, Amber, WISP, 15x45mm, GPI 13-425
SDP
Catalog No. 50230781
Encompass_Preferred
Vial, 4.0mL, Amber, WISP, 15x45mm, GPI 13-425. This vial is designed to work in Shimadzu and Waters WISP™ 48 position auto samplers. Amber borosilicate. These vials incorporate the unique Step Vial design that precisely centers a limited volume insert in the neck of the vial. Available with graduated marking spots.
Catalog No. 50-230-781 Supplier Chemglass Life Sciences Supplier No. CV12571545
Vial, 4.0mL, Amber, WISP, 15x45mm, GPI 13-425. This vial is designed to work in Shimadzu and Waters WISP™ 48 position auto samplers. Amber borosilicate. These vials incorporate the unique Step Vial design that precisely centers a limited volume insert in the neck of the vial. Available with graduated marking spots.