Learn More
Chemglass Life Sciences Vial, 4.0mL, Amber with White Grad Spot, WISP, 15x45mm, GPI 13-425
SDP
Catalog No. 501532466
Encompass_Preferred
Vial, 4.0mL, Amber with White Grad Spot, WISP, 15x45mm, GPI 13-425. This vial is designed to work in Shimadzu and Waters WISP 48 position auto samplers. These vials incorporate the unique Step Vial design that precisely centers a limited volume insert in the neck of the vial.
Catalog No. 50-153-2466 Supplier Chemglass Life Sciences Supplier No. CV12591545
Vial, 4.0mL, Amber with White Grad Spot, WISP, 15x45mm, GPI 13-425. This vial is designed to work in Shimadzu and Waters WISP 48 position auto samplers. These vials incorporate the unique Step Vial design that precisely centers a limited volume insert in the neck of the vial.