Learn More
Chemglass Life Sciences Vial, 4.0mL, Clear, WISP, 15x45mm, GPI 13-425
SDP
Catalog No. 50230780
Encompass_Preferred
Vial, 4.0mL, Clear, WISP, 15x45mm, GPI 13-425. This vial is designed to work in Shimadzu and Waters WISP™ 48 position auto samplers. Clear. These vials incorporate the unique Step Vial design that precisely centers a limited volume insert in the neck of the vial. Available with graduated marking spots.
Catalog No. 50-230-780 Supplier Chemglass Life Sciences Supplier No. CV12561545
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
View Alternate Product
Vial, 4.0mL, Clear, WISP, 15x45mm, GPI 13-425. This vial is designed to work in Shimadzu and Waters WISP™ 48 position auto samplers. Clear. These vials incorporate the unique Step Vial design that precisely centers a limited volume insert in the neck of the vial. Available with graduated marking spots.