Learn More
Chemglass Life Sciences Vial, 4.0mL, Clear with White Grad Spot, WISP, 15x45mm, GPI 13-425
SDP
Catalog No. 5000980651
Encompass_Preferred
This vial is designed to work in Shimadzu and Waters WISP™ 48 position auto samplers. Choose from clear or amber borosilicate. These vials incorporate the unique Step Vial design that precisely centers a limited volume insert in the neck of the vial. Available with graduated marking spots.
Catalog No. 50-009-80651 Supplier Chemglass Life Sciences Supplier No. CV12581545
This vial is designed to work in Shimadzu and Waters WISP™ 48 position auto samplers. Choose from clear or amber borosilicate. These vials incorporate the unique Step Vial design that precisely centers a limited volume insert in the neck of the vial. Available with graduated marking spots.