Learn More
Chemglass Life Sciences Vial, 4.0mL, Polypropylene, Shell, 15x45mm
SDP
Catalog No. 501532506
Encompass_Preferred
Vial, 4.0mL, Polypropylene, Shell, 15x45mm. Shell vials are the most economical alternative for use in an auto sampler. Feature thicker walls for safer sample handling.
Catalog No. 50-153-2506 Supplier Chemglass Life Sciences Supplier No. CV21171545
Vial, 4.0mL, Polypropylene, Shell, 15x45mm. Shell vials are the most economical alternative for use in an auto sampler. Feature thicker walls for safer sample handling.