Learn More
Med Vet International Blue Prescription Vial, 40 Dram, 200/Case
Supplier:  Med Vet International VIAL40DB
Encompass_Preferred
The Oasis 40 Dram child resistant prescription pill bottle is designed for storing medication. This prescription bottle comes in Amber and has a push down and turn cap making it difficult for children to open, but easy for the elderly. The light resistant vial is child resistant with 5% free easy open snap on caps. This product contains 200/case prescription vials. Height without cap: 9.5cm (3.74") Diameter 4.5cm (1.77"). EASY TO USE:  These bottles are easy to use and you can safely store your medicine or other small items in them. These prescription pharmacy pill bottles have push and turn caps. PERFECT TO ORGANIZE AND STORE PILLS:  These medicine containers are perfect to organize and store pills. The bottles can be used for pharmacies, physicians, labs, hospitals, and vets. LIGHTWEIGHT AND DURABLE PLASTIC:  These medicine bottles are made of premium quality plastic material.
Catalog No. 50-264-3420