Learn More
Industrial Glassware VIAL 40ML AMBER CLOSED-TOP
Catalog No. NC2388388
Encompass_Preferred
Manufactured from borosilicate glass, these vials provide excellent chemical-resistant storage.Pre-cleanedAdhesive-free 24-400 closed-top Teflon-lined cap.Pre-cleaned vials are processed under strict quality-control conditions. Certificate of Analysis available at no additional charge.
Catalog No. NC2388388 Supplier Industrial Glassware Supplier No. 2795ATPC
Manufactured from borosilicate glass, these vials provide excellent chemical-resistant storage.Pre-cleanedAdhesive-free 24-400 closed-top Teflon-lined cap.Pre-cleaned vials are processed under strict quality-control conditions. Certificate of Analysis available at no additional charge.