Learn More
Ace Glass, Inc. 40mL (10 dram) amber EPA vials with 24-400 open top, white polypropylene caps with PTFE/Silicone liners, 28x96mm, cs/72, Wheaton 225315
SDP
Catalog No. 50976421
Encompass
40mL (10 dram) amber EPA vials with 24-400 open top, white polypropylene caps with PTFE/Silicone liners, 28x96mm, cs/72, Wheaton 225315
Catalog No. 50-976-421 Supplier Ace Glass, Inc. Supplier No. 878125
40mL (10 dram) amber EPA vials with 24-400 open top, white polypropylene caps with PTFE/Silicone liners, 28x96mm, cs/72, Wheaton 225315