Learn More
Chemglass Life Sciences Vial, 6.0mL, Clear, 20mm Headspace, Round Bottom, 22x38mm, 20mm Crimp, for use with Perkin Elmer Autosamplers
SDP
Catalog No. 501532469
Encompass_Preferred
Vial, 6.0mL, Clear, 20mm Headspace, Round Bottom, 22x38mm, 20mm Crimp, for use with Perkin Elmer Autosamplers. These vials are manufactured from Type 1 Class A Borosilicate glass. Please Note: Pressure Release Aluminum Seals are Designed to Open When the Internal Pressure Exceeds 3.0 +/- 0.5 Bar.
Catalog No. 50-153-2469 Supplier Chemglass Life Sciences Supplier No. CV12812238
Vial, 6.0mL, Clear, 20mm Headspace, Round Bottom, 22x38mm, 20mm Crimp, for use with Perkin Elmer Autosamplers. These vials are manufactured from Type 1 Class A Borosilicate glass. Please Note: Pressure Release Aluminum Seals are Designed to Open When the Internal Pressure Exceeds 3.0 +/- 0.5 Bar.