Learn More
Med Vet International Amber Prescription Vial, 6 Dram, 600/Case
Catalog No. 502643414
Encompass_Preferred
The Oasis 6 Dram child resistant prescription pill bottle is designed for storing medication. This prescription bottle comes in Amber and has a push down and turn cap making it difficult for children to open, but easy for the elderly. The light resistant vial is child resistant with 5% free easy open snap on caps. This product contains 600/case prescription vials. Height without cap: 6.3cm (2.48") Diameter 2.2cm (0.87"). EASY TO USE:  These bottles are easy to use and you can safely store your medicine or other small items in them. These prescription pharmacy pill bottles have push and turn caps. PERFECT TO ORGANIZE AND STORE PILLS:  These medicine containers are perfect to organize and store pills. The bottles can be used for pharmacies, physicians, labs, hospitals, and vets.
Catalog No. 50-264-3414 Supplier Med Vet International Supplier No. VIAL6D
The Oasis 6 Dram child resistant prescription pill bottle is designed for storing medication. This prescription bottle comes in Amber and has a push down and turn cap making it difficult for children to open, but easy for the elderly. The light resistant vial is child resistant with 5% free easy open snap on caps. This product contains 600/case prescription vials. Height without cap: 6.3cm (2.48") Diameter 2.2cm (0.87"). EASY TO USE:  These bottles are easy to use and you can safely store your medicine or other small items in them. These prescription pharmacy pill bottles have push and turn caps. PERFECT TO ORGANIZE AND STORE PILLS:  These medicine containers are perfect to organize and store pills. The bottles can be used for pharmacies, physicians, labs, hospitals, and vets.