Learn More
Cole-Parmer Headspace Vial, Amber Glass, 18 mm, Precision Thread Screw Neck, Round Bottom, 10 mL; 1000/pk
Catalog No. NC1504311 Shop All Cole-Parmer Products
Encompass_Preferred
Type I Class 51 borosilicate glass provide excellent chemical resistance to various solutions. Vial design ensures full automation compatibility for headspace analyzers for both handling and needle positioning. Crimp and screw caps are available with various septum materials.
Catalog No. NC1504311 Supplier Cole-Parmer Supplier No. 9870497
Type I Class 51 borosilicate glass provide excellent chemical resistance to various solutions. Vial design ensures full automation compatibility for headspace analyzers for both handling and needle positioning. Crimp and screw caps are available with various septum materials.