Learn More
Chemglass Life Sciences Storage Vial, Amber, 40mL, 28 x 95mm, 24-414 White Solid Cap, Cell Pack
SDP
Catalog No. 50143878
Encompass_Preferred
Storage Vial, Amber, 40mL, 28 x 95mm, 24-414 White Solid Cap, Cell Pack. For storage applications, these vials are packaged in a SBS coated white tray complete with SBS partition. Boxes are marked with alpha-numeric coordinates for fast and easy vial identification. Caps are supplied installed on the vials and all sizes have the TFE lined silicone SURE-LINK septa.
Catalog No. 50-143-878 Supplier Chemglass Life Sciences Supplier No. CG4908A04
Storage Vial, Amber, 40mL, 28 x 95mm, 24-414 White Solid Cap, Cell Pack. For storage applications, these vials are packaged in a SBS coated white tray complete with SBS partition. Boxes are marked with alpha-numeric coordinates for fast and easy vial identification. Caps are supplied installed on the vials and all sizes have the TFE lined silicone SURE-LINK septa.