Learn More
Chemglass Life Sciences Storage Vial, Amber, 4mL, 15 x 45mm, 13-425 Green Solid Cap, Cell Pack
SDP
Catalog No. 50143877
Encompass_Preferred
Storage Vial, Amber, 4mL, 15 x 45mm, 13-425 Green Solid Cap, Cell Pack. For storage applications, these vials are packaged in a SBS coated white tray complete with SBS partition. Boxes are marked with alpha-numeric coordinates for fast and easy vial identification. Caps are supplied installed on the vials and all sizes have the TFE lined silicone SURE-LINK septa. The 4mL is a 1 dram.
Catalog No. 50-143-877 Supplier Chemglass Life Sciences Supplier No. CG4908A01
Storage Vial, Amber, 4mL, 15 x 45mm, 13-425 Green Solid Cap, Cell Pack. For storage applications, these vials are packaged in a SBS coated white tray complete with SBS partition. Boxes are marked with alpha-numeric coordinates for fast and easy vial identification. Caps are supplied installed on the vials and all sizes have the TFE lined silicone SURE-LINK septa. The 4mL is a 1 dram.