Learn More
Chemglass Life Sciences Storage Vial, Amber, 8mL, 17 x 60mm, 15-425 Green Solid Cap, Cell Pack
SDP
Catalog No. 50143869
Encompass_Preferred
Storage Vial, Amber, 8mL, 17 x 60mm, 15-425 Green Solid Cap, Cell Pack. For storage applications, these vials are packaged in a SBS coated white tray complete with SBS partition. Boxes are marked with alpha-numeric coordinates for fast and easy vial identification. Caps are supplied installed on the vials and all sizes have the TFE lined silicone SURE-LINK septa. The 8mL is a 2 dram.
Catalog No. 50-143-869 Supplier Chemglass Life Sciences Supplier No. CG4908A02
Storage Vial, Amber, 8mL, 17 x 60mm, 15-425 Green Solid Cap, Cell Pack. For storage applications, these vials are packaged in a SBS coated white tray complete with SBS partition. Boxes are marked with alpha-numeric coordinates for fast and easy vial identification. Caps are supplied installed on the vials and all sizes have the TFE lined silicone SURE-LINK septa. The 8mL is a 2 dram.