Learn More
Microsolv Technology Corporation Vial Aq Screw Top Clear Glass
Catalog No. 5001452287
Encompass_Preferred
Vials, Screw Top, Glass. Clear, 2ml, 9-425mm threads, 12x32mm outer dimensions. For use as an autosampler vial or storage bottle. AQ Brand. 100/PK.
Catalog No. 50-014-52287 Supplier Microsolv Technology Corporation Supplier No. 9509S0CVAQ
Vials, Screw Top, Glass. Clear, 2ml, 9-425mm threads, 12x32mm outer dimensions. For use as an autosampler vial or storage bottle. AQ Brand. 100/PK.