Learn More
Microsolv Technology Corporation Vial Aqr Screw Top Amber Wo
Catalog No. 5001452303
Encompass_Preferred
Vials, Screw Top, Glass. Amber, 2ml, with Write-On Patch and fill lines, 9-425mm threads, 12x32mm outer dimensions. For use as an autosampler vial. RSA Brand. 100/PK.
Catalog No. 50-014-52303 Supplier Microsolv Technology Corporation Supplier No. 9509SWAVRS
Vials, Screw Top, Glass. Amber, 2ml, with Write-On Patch and fill lines, 9-425mm threads, 12x32mm outer dimensions. For use as an autosampler vial. RSA Brand. 100/PK.