Learn More
Microsolv Technology Corporation Vial Aqr Screw Top Clear Wo
Catalog No. 5001452305
Encompass_Preferred
Vials, Screw Top, Glass. Clear, 2ml, with Write-On Patch and fill lines, 9-425mm threads, 12x32mm outer dimensions. For use as an autosampler vial. RSA Brand. 100/PK.
Catalog No. 50-014-52305 Supplier Microsolv Technology Corporation Supplier No. 9509SWCVRS
Vials, Screw Top, Glass. Clear, 2ml, with Write-On Patch and fill lines, 9-425mm threads, 12x32mm outer dimensions. For use as an autosampler vial. RSA Brand. 100/PK.