Learn More
Cole-Parmer Kinesis Polypropylene Storage Box, Blue, 25 x 5-, 10-, or 20-mL Vials; Each
Catalog No. NC2143498 Shop All Cole-Parmer Products
Encompass_Preferred
Sturdy polypropylene storage boxes hold vials in placeInspect, reference, and identify samples through the and clear lid and easy-to-read numeric gridSelect models with alphanumeric coding to help organize and keep track of samplesDurable polypropylene storage boxes are available in a range of colors for easy identification. Choose from boxes that accommodate different tube sizes for any application.
This item is not returnable. View return policy
Catalog No. NC2143498 Supplier Cole-Parmer Supplier No. 9870260
Sturdy polypropylene storage boxes hold vials in placeInspect, reference, and identify samples through the and clear lid and easy-to-read numeric gridSelect models with alphanumeric coding to help organize and keep track of samplesDurable polypropylene storage boxes are available in a range of colors for easy identification. Choose from boxes that accommodate different tube sizes for any application.