Learn More
Electron Microscopy Sciences Vial Caddy 48 Place 23 mm
SDP
Catalog No. 50949624
Encompass_Preferred
Heavy-duty vial caddy protects up to 48 of the popular 23 mm diameter glass vials. Machined from a solid block of 1.25" HDPE sheet offering complete protection during storage and transport. The caddy is supplied with a snug fitting cover and non-skid feet. It is white in color.
Catalog No. 50-949-624 Supplier Electron Microscopy Sciences Supplier No. 72642
Heavy-duty vial caddy protects up to 48 of the popular 23 mm diameter glass vials. Machined from a solid block of 1.25" HDPE sheet offering complete protection during storage and transport. The caddy is supplied with a snug fitting cover and non-skid feet. It is white in color.