Learn More
Microsolv Technology Corporation Vial/Cap 1Ml Clear 96Pos
Catalog No. 5001451811
Encompass_Preferred
Vial & Cap Kit includes 250 EA 1ml, Push Top, Clear, 8x40mm, Autosampler Vials and 250 EA matching Push Top Caps. AQ Brand. 250/PK.
Catalog No. 50-014-51811 Supplier Microsolv Technology Corporation Supplier No. 950010CP
Vial & Cap Kit includes 250 EA 1ml, Push Top, Clear, 8x40mm, Autosampler Vials and 250 EA matching Push Top Caps. AQ Brand. 250/PK.