Learn More
Microsolv Technology Corporation Vial & Cap Kit includes 100 EA 300ul, Snap Top, Clear polypropylene LCMS Compatible Autosampler Vials and 100 EA matching Snap Caps with Silicone Rubber / PTFE Septa fitted in the Blue Caps. 1/PK.
Catalog No. 502270130
Encompass_Preferred
Easy Purchase Packs are a convenient way to order and inventory autosampler vials with their matching caps. The kit packaging will open and re-close making it useful for keeping vial and cap types matched, organized and clean during use. This is one of our Easy Purchase Packs.
This item is not returnable. View return policy
Catalog No. 50-227-0130 Supplier Microsolv Technology Corporation Supplier No. 9532C1PC
Easy Purchase Packs are a convenient way to order and inventory autosampler vials with their matching caps. The kit packaging will open and re-close making it useful for keeping vial and cap types matched, organized and clean during use. This is one of our Easy Purchase Packs.