Learn More
Microsolv Technology Corporation Vial & Cap Kit Includes 1000 EA 300ul, Screw Top, Clear polypropylene Autosampler Vials and 1000 EA matching Screw Caps with Silicone Rubber / PTFE Septa, fitted in the Robo Style, Blue Caps. 1/PK.
Catalog No. 502270145
Encompass_Preferred
Easy Purchase Packs are a convenient way to order and inventory autosampler vials with their matching caps.
This item is not returnable. View return policy
Catalog No. 50-227-0145 Supplier Microsolv Technology Corporation Supplier No. 9532S0P05CK
Easy Purchase Packs are a convenient way to order and inventory autosampler vials with their matching caps.