Learn More
Microsolv Technology Corporation Vial & Cap Kit Includes 100 EA 2ml, Screw Top, Amber Autosampler Vials with Write-On Patch and fill lines and 100 EA matching Screw Caps with Tan Silicone Rubber / White PTFE, Pre-Slit Septa bonded in the Blue Caps. Vials and Caps are Pre-assembled. 1/PK.
Catalog No. 502270321
Encompass_Preferred
Easy Purchase Packs are a convenient way to order and inventory autosampler vials with their matching caps.
This item is not returnable. View return policy
Catalog No. 50-227-0321 Supplier Microsolv Technology Corporation Supplier No. 9509S3WAPAQA
Easy Purchase Packs are a convenient way to order and inventory autosampler vials with their matching caps.